Learn More
Aviva Systems Biology Corp PLEKHG2 Antibody - middle region (ARP59650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PLEKHG2. Alternate names: CLG, DKFZp667J2325, FLJ00018, FLJ22458, FLJ38638, ARHGEF42ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-767