Learn More
Aviva Systems Biology Corp PLEKHH2 Antibody - C-terminal region (ARP53218_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PLEKHH2. Alternate names: KIAA2028, PLEKHH1LApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53702