Learn More
Aviva Systems Biology Corp PLEKHJ1 Antibody - C-terminal region (ARP62828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHJ1. It was validated on Western Blot. . Alternate names: FLJ10297, GNRPXApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-287-780