Learn More
Aviva Systems Biology Corp PLEKHO1 Antibody - N-terminal region (ARP47817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHO1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PLEKHO1. Alternate names: CKIP-1, OC120, RP11-458I7.3ApplicationWB. Species reactivity: Cow, Human, Pig, Rat
Catalog No. 50-285-330