Learn More
OriGene PLENTI-EF1A-C-MGFP 10UG
SDP
Catalog No. NC1724558
Encompass_Preferred
pLenti-EF1a-C-mGFP, Lenti Vector with EF1 alpha promoter and C-terminal mGFP tag, 10ug
Catalog No. NC1724558 Supplier OriGene Supplier No. PS100084
pLenti-EF1a-C-mGFP, Lenti Vector with EF1 alpha promoter and C-terminal mGFP tag, 10ug