Learn More
Medchemexpress LLC HY-10046 100mg , Plerixafor AMD3100;JM3100 CAS:110078-46-1 Purity:98%
SDP
Catalog No. 501492697
Encompass_Preferred
Medchemexpress, HY-10046 100mg Plerixafor AMD3100;JM3100 CAS:110078-46-1 Formula:C28H54N8 IC50: 44 nM (CXCR4) Purity:98% Plerixafor is a selective CXCR4 antagonist with IC 50 of 44 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2697 Supplier Medchemexpress LLC Supplier No. HY10046100MG
Medchemexpress, HY-10046 100mg Plerixafor AMD3100;JM3100 CAS:110078-46-1 Formula:C28H54N8 IC50: 44 nM (CXCR4) Purity:98% Plerixafor is a selective CXCR4 antagonist with IC 50 of 44 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.