Learn More
Apexbio Technology LLC Plerixafor (AMD3100), 25mg. Cas: 110078-46-1 MFCD: MFCD05662218. CXCR4 chemokine receptor antagonist
SDP
Supplier:  Apexbio Technology LLC A202525
Encompass_Preferred
Plerixafor (AMD3100) is a small-molecule antagonist of CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5.7 nM, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4730