Learn More
Cayman Chemical PlerIxafor-d4 1mg
SDP
Catalog No. 502934016
Encompass_Preferred
An internal standard for the quantification of plerixafor by GC- or LC-MS
Catalog No. 50-293-4016 Supplier Cayman Chemical Supplier No. 264901 MG
An internal standard for the quantification of plerixafor by GC- or LC-MS