Learn More
Cayman Chemical Plerixafor hydrochloride hyd
SDP
Catalog No. 502887129
Encompass_Preferred
An antagonist of CXCR4 (IC50s = 0.02-0.13 UG/ml); selective for CXCR4 over CXCR1-3 and CXCR5-9 (IC50s = >25 UG/ml); decreases infectious virus content in the supernatant of Jurkat cells chronically infected with HIV-1(IIIB) (EC50 = ~0.02 UG/ml); mobilizes murine and human hematopoietic stem and murine long-term repopulating cells for transplantation; increases T cell trafficking in mouse blood, spleen, and central nervous system; decreases the number of 4T1 murine mammary carcinoma cells in the lung in a mouse model of lung metastasis at 1.25 mg/kg twice per day
Catalog No. 50-288-7129 Supplier Cayman Chemical Supplier No. 100113321 MG
An antagonist of CXCR4 (IC50s = 0.02-0.13 UG/ml); selective for CXCR4 over CXCR1-3 and CXCR5-9 (IC50s = >25 UG/ml); decreases infectious virus content in the supernatant of Jurkat cells chronically infected with HIV-1(IIIB) (EC50 = ~0.02 UG/ml); mobilizes murine and human hematopoietic stem and murine long-term repopulating cells for transplantation; increases T cell trafficking in mouse blood, spleen, and central nervous system; decreases the number of 4T1 murine mammary carcinoma cells in the lung in a mouse model of lung metastasis at 1.25 mg/kg twice per day