Learn More
Cayman Chemical PleuromutIlIn 10mg
SDP
Catalog No. 502985368
Encompass_Preferred
An antibiotic that inhibits bacterial protein synthesis by binding to bacterial ribosomes in the peptidyl transferase component of the 50S subunit and inhibiting peptide bond formation
Catalog No. 50-298-5368 Supplier Cayman Chemical Supplier No. 1945210 MG
An antibiotic that inhibits bacterial protein synthesis by binding to bacterial ribosomes in the peptidyl transferase component of the 50S subunit and inhibiting peptide bond formation