Learn More
Apexbio Technology LLC Pleuromutilin 125-65-5 5mg
SDP
Catalog No. 502240747
Encompass_Preferred
Pleuromutilin (125-65-5) is a small-molecule inhibitor targeting the bacterial ribosome 50S subunit It is designed to inhibit bacterial protein synthesis by selectively binding to the peptidyl-transferase center thereby suppressing peptide bond formation Pleuromutilin exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies Pleuromutilin demonstrates antibacterial activity with reported IC50 values typically in the low micromolar and sub-micromolar ranges Based on these pharmacological properties Pleuromutilin holds research potential in applications such as treatment of Gram-positive bacterial infections and as a chemical scaffold for developing veterinary and topical antibacterial agents
This item is not returnable. View return policy
Catalog No. 50-224-0747 Supplier Apexbio Technology LLC Supplier No. C55435
Pleuromutilin (125-65-5) is a small-molecule inhibitor targeting the bacterial ribosome 50S subunit It is designed to inhibit bacterial protein synthesis by selectively binding to the peptidyl-transferase center thereby suppressing peptide bond formation Pleuromutilin exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies Pleuromutilin demonstrates antibacterial activity with reported IC50 values typically in the low micromolar and sub-micromolar ranges Based on these pharmacological properties Pleuromutilin holds research potential in applications such as treatment of Gram-positive bacterial infections and as a chemical scaffold for developing veterinary and topical antibacterial agents