Learn More
Electron Microscopy Sciences Plexiglass Microscope Slides
SDP
Catalog No. 501926620
Encompass_Preferred
A Fool-Proof method for the mounting of serial sections on Single Hole Grids 1 These plexiglass slides are 3x1 inches (75-25 mm) and 1 mm thick. They prevent wrinkling and section loss while mounting sections on the grid.
Catalog No. 50-192-6620 Supplier Electron Microscopy Sciences Supplier No. 7189110
A Fool-Proof method for the mounting of serial sections on Single Hole Grids 1 These plexiglass slides are 3x1 inches (75-25 mm) and 1 mm thick. They prevent wrinkling and section loss while mounting sections on the grid.