Learn More
ECM Bioscience Plexin A1, B1, and D1 Receptors Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068780
Encompass_Preferred
The Plexin Receptor kit can be used to monitor the expression of Plexin A1, Plexin B1, and Plexin D1. The kit includes three antibodies for detecting different regions of Plexin D1, two antibodies to different regions in the Sema domain of Plexin A1, and an antibody that detects the C-terminal region of Plexin B1. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8780 Supplier ECM Bioscience Supplier No. PK6580
The Plexin Receptor kit can be used to monitor the expression of Plexin A1, Plexin B1, and Plexin D1. The kit includes three antibodies for detecting different regions of Plexin D1, two antibodies to different regions in the Sema domain of Plexin A1, and an antibody that detects the C-terminal region of Plexin B1. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms