Learn More
ORS Nasco PLG LLIT CRD 100PR/PK 10PK/CS
Catalog No. 502551614
Encompass_Preferred
Honeywell part# LL-30. Honeywell Laser Lite Ear Plugs Corded: Corded Disposable: Yes Metal Detectable: No Noise Reduction Rating: 32 dB Plug Shape: T-Shape Size: Universal ANSI S3.1974: Yes Cord Color: Yellow Cord Material: Vinyl CSA Z94.2: Class AL High Visibility: Yes Poly bag
Catalog No. 50-255-1614 Supplier ORS Nasco Supplier No. 154LL30
Honeywell part# LL-30. Honeywell Laser Lite Ear Plugs Corded: Corded Disposable: Yes Metal Detectable: No Noise Reduction Rating: 32 dB Plug Shape: T-Shape Size: Universal ANSI S3.1974: Yes Cord Color: Yellow Cord Material: Vinyl CSA Z94.2: Class AL High Visibility: Yes Poly bag