Learn More
Medchemexpress LLC HY-B2247 500mg , PLGA (50:50) CAS:34346-01-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2247500MG
Encompass_Preferred
HY-B2247 500mg PLGA (50:50) CAS: 34346-01-5 PLGA (poly(lactic-co-glycolic acid)) (50:50) is a copolymer of poly lactic acid (PLA) and poly glycolic acid (PGA) which can be used to fabricate devices for drug delivery and tissue engineering applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0302