Learn More
NANOSOFT POLYMERS PLGA-Amine/PLGA-NH2 (85:15) MW 20K Size 1g
Catalog No. NC3666409
Encompass
Amine capped poly(D,L-lactide-co-glycolide) (PLGA-Amine, PLGA-NH2, NH2-PLGA, D,L-LA/GA: 85:15) can be used to prepare nanoparticles, which can be further modified by amine-reactive entities. PLGA-amine can be used to prepare polymer-drug conjugates.Poly (lactic-co-glycolic acid) (PLGA) is one of the most effective biodegradable polymeric nanoparticles (NPs). It has been approved by the US FDA to use in drug delivery systems due to controlled and sustained- release properties, low toxicity, and biocompatibility with tissue and cells.
Catalog No. NC3666409 Supplier NANOSOFT POLYMERS Supplier No. 1242920K1G
Amine capped poly(D,L-lactide-co-glycolide) (PLGA-Amine, PLGA-NH2, NH2-PLGA, D,L-LA/GA: 85:15) can be used to prepare nanoparticles, which can be further modified by amine-reactive entities. PLGA-amine can be used to prepare polymer-drug conjugates.Poly (lactic-co-glycolic acid) (PLGA) is one of the most effective biodegradable polymeric nanoparticles (NPs). It has been approved by the US FDA to use in drug delivery systems due to controlled and sustained- release properties, low toxicity, and biocompatibility with tissue and cells.