Learn More
NANOSOFT POLYMERS PLGA-PEG-BIO | 100mg
Catalog No. NC1649862
Encompass
PLGA-PEG-BIO | PLGA MW: 10000 | PEG MW: 5000 | 100mg
Catalog No. NC1649862 Supplier NANOSOFT POLYMERS Supplier No. PLGAPEGBIO100MG
PLGA-PEG-BIO | PLGA MW: 10000 | PEG MW: 5000 | 100mg