Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Plga-peg-mal (20kda-5.0kda, la:ga ratio 40:60) | 95.0% | 20kDA to 5.0kDA | 5 MG
SDP

Catalog No. 5000360150
Encompass_Preferred

PLGA-PEG-MAL (20kDA-5.0kDA, LA:GA ratio 40:60) is a type of poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticles. It has a molecular weight range of 20kDA to 5.0kDA and contains a 40:60 ratio of lactic acid (LA) to glycolic acid (GA) molecules. This solid, white to off-white compound has a purity of 95.0% and is intended for research use only.

  • Type of poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticles
  • Contains a 40:60 ratio of lactic acid (LA) to glycolic acid (GA) molecules
  • Molecular weight of 20kDA to 5.0kDA
  • Purity of 95.0%
  • Solid, white to off-white appearance
  • For research use only

Catalog No. 50-003-60150 Supplier Medchemexpress LLC Supplier No. HY148775B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PLGA-PEG-MAL (20kDA-5.0kDA, LA:GA ratio 40:60) is a type of poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticles. It has a molecular weight range of 20kDA to 5.0kDA and contains a 40:60 ratio of lactic acid (LA) to glycolic acid (GA) molecules. This solid, white to off-white compound has a purity of 95.0% and is intended for research use only.

  • Type of poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticles
  • Contains a 40:60 ratio of lactic acid (LA) to glycolic acid (GA) molecules
  • Molecular weight of 20kDA to 5.0kDA
  • Purity of 95.0%
  • Solid, white to off-white appearance
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.