Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PLGA-PEG-MAL (60kDA-3.4kDA, LA:GA ratio 75:25) | 95.0% | 60kDA to 3.4kDA | 5 MG
SDP

Catalog No. 5000362043
Encompass_Preferred

PLGA-PEG-MAL (60kDA-3.4kDA, LA:GA ratio 75:25) is a type of poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticle. It has a molecular weight ranging from 60kDA to 3.4kDA and contains a 75:25 ratio of lactic acid (LA) to glycolic acid (GA) molecules. This product is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Purity: 95.0%
  • Solubility in DMSO: 100 mg/mL (requires sonication; use newly opened DMSO)
  • Classified as oligonucleotides, polymers, biochemical assay reagents, and drug delivery agents

Catalog No. 50-003-62043 Supplier Medchemexpress LLC Supplier No. HY1487755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PLGA-PEG-MAL (60kDA-3.4kDA, LA:GA ratio 75:25) is a type of poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticle. It has a molecular weight ranging from 60kDA to 3.4kDA and contains a 75:25 ratio of lactic acid (LA) to glycolic acid (GA) molecules. This product is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Purity: 95.0%
  • Solubility in DMSO: 100 mg/mL (requires sonication; use newly opened DMSO)
  • Classified as oligonucleotides, polymers, biochemical assay reagents, and drug delivery agents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.