Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PLGA12000-PEG2000-NH2 | 98.0% | 100 MG
SDP

Catalog No. 5000354322
Encompass_Preferred

PLGA-PEG-NH2 is a material used to synthesize nanomicelles. PLGA-PEG-NH2 nanomicelle serves as an efficient delivery system for Irinotecan, targeting colorectal cancer and hepatocellular carcinoma.

  • Material to synthesize nanomicelles
  • Efficient delivery system for Irinotecan
  • Targets colorectal cancer
  • Targets hepatocellular carcinoma
  • Appearance: solid
  • Color: white to light yellow
  • Purity: 98.0%
  • Store at -20°C, under nitrogen
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-003-54322 Supplier Medchemexpress LLC Supplier No. HY145396100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PLGA-PEG-NH2 is a material used to synthesize nanomicelles. PLGA-PEG-NH2 nanomicelle serves as an efficient delivery system for Irinotecan, targeting colorectal cancer and hepatocellular carcinoma.

  • Material to synthesize nanomicelles
  • Efficient delivery system for Irinotecan
  • Targets colorectal cancer
  • Targets hepatocellular carcinoma
  • Appearance: solid
  • Color: white to light yellow
  • Purity: 98.0%
  • Store at -20°C, under nitrogen
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.