Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse PLGF-2 (PGF) ELISA Kit

Catalog No. EMPGF
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMPGF 96 Tests
EMPGFX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EMPGF Supplier Invitrogen™ Supplier No. EMPGF
Add to Cart
Add to Cart

ELISA

The Mouse PLGF-2 (PGF) ELISA quantitates Ms PLGF-2 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms PLGF-2. Principle of the method The Mouse PLGF-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49764
Assay Range 1-400 pg/mL
Assay Sensitivity 1 pg/mL
Calibration Range 1 pg/mL to 400 pg/mL
Content And Storage Pre-coated 96-well plate, standard, detector antibody, streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers. May be stored at 2–8°C for up to 6 months from the date of shipment.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity The sandwich ELISA antibody pair detects Mouse PLGF-2.
Sufficient For 96 Assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760;
Gene ID (Entrez) 18654
Gene Symbol PGF
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases PlGF
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym AI854365, PIGF, Plgf
Target PGF
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.