Learn More
ACROBiosystems ACROBiosystems Rhesus macaque PLGF / PGF Protein, His Tag
Catalog No. 501957212
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque PLGF, His Tag (PGF-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession # F7HB10-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 28-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) at 5 μg/mL (100 μL/well) can bind Rhesus macaque PLGF, His Tag (Cat. No. PGF-R52H0) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-195-7212 Supplier ACROBiosystems Supplier No. PGFR52H01MG
Recombinant Protein;1MG;Rhesus macaque PLGF, His Tag (PGF-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession # F7HB10-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 28-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) at 5 μg/mL (100 μL/well) can bind Rhesus macaque PLGF, His Tag (Cat. No. PGF-R52H0) with a linear range of 2-40 ng/mL (QC tested).