Learn More
Medchemexpress LLC PLGF Human HEK293 10ug
SDP
Catalog No. 5000185288
Encompass_Preferred
Recombinant human PLGF-2 produced in HEK293 cells with a polyhistidine tag, suitable for research on angiogenesis and endothelial cell biology. The protein is active as a disulfide-linked homodimer, interacts with VEGFR-1/FLT1 and neuropilins, and is supplied with endotoxin control; follow the manufacturer datasheet for reconstitution and handling instructions.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Includes a His affinity tag to facilitate purification and detection.
  • Active as a disulfide-linked homodimer that interacts with VEGFR-1 and neuropilins.
  • Low endotoxin level suitable for cell-based assays.
  • Stable when stored frozen; aliquot after reconstitution to preserve activity.
Catalog No. 50-001-85288 Supplier Medchemexpress LLC Supplier No. HYP7074910UG
Recombinant human PLGF-2 produced in HEK293 cells with a polyhistidine tag, suitable for research on angiogenesis and endothelial cell biology. The protein is active as a disulfide-linked homodimer, interacts with VEGFR-1/FLT1 and neuropilins, and is supplied with endotoxin control; follow the manufacturer datasheet for reconstitution and handling instructions.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Includes a His affinity tag to facilitate purification and detection.
  • Active as a disulfide-linked homodimer that interacts with VEGFR-1 and neuropilins.
  • Low endotoxin level suitable for cell-based assays.
  • Stable when stored frozen; aliquot after reconstitution to preserve activity.