Learn More
Medchemexpress LLC PLGF Human HEK293 50ug
SDP
Catalog No. 5000185327
Encompass_Preferred
Recombinant human PLGF-2 produced in HEK293 cells and provided as a lyophilized, His-tagged protein for research applications in angiogenesis and endothelial cell biology. Supplied in small research-scale quantities, the protein is formulated to preserve stability during storage and handling and includes guidance for reconstitution and long-term storage to maintain activity.
  • Mammalian expression preserves native post-translational modifications.
  • Lyophilized format enables room temperature shipment and extended shelf life.
  • His tag facilitates detection and affinity purification when needed.
  • Formulated in buffered saline for compatibility with common assays.
  • Storage and reconstitution guidance included to help retain bioactivity.
Catalog No. 50-001-85327 Supplier Medchemexpress LLC Supplier No. HYP7074950UG
Recombinant human PLGF-2 produced in HEK293 cells and provided as a lyophilized, His-tagged protein for research applications in angiogenesis and endothelial cell biology. Supplied in small research-scale quantities, the protein is formulated to preserve stability during storage and handling and includes guidance for reconstitution and long-term storage to maintain activity.
  • Mammalian expression preserves native post-translational modifications.
  • Lyophilized format enables room temperature shipment and extended shelf life.
  • His tag facilitates detection and affinity purification when needed.
  • Formulated in buffered saline for compatibility with common assays.
  • Storage and reconstitution guidance included to help retain bioactivity.