Learn More
Medchemexpress LLC PLGF Mouse HEK293 10ug
SDP
Catalog No. 5000184702
Encompass_Preferred
Recombinant mouse placental growth factor (PLGF) expressed in HEK293 cells and provided lyophilized with a C-terminal His tag. PLGF binds VEGFR-1/FLT1 to stimulate angiogenesis and endothelial cell proliferation and migration; structurally it forms antiparallel homodimers and can heterodimerize with VEGF family members. Reconstitute according to the manufacturer instructions and aliquot for long-term storage.
  • Recombinant mouse PLGF expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Promotes angiogenesis via VEGFR-1/FLT1 binding.
  • Antiparallel homodimer that can form heterodimers with VEGF family members.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Approximate molecular weight 27-30 kDa by SDS-PAGE.
  • Store lyophilized at -20°C; aliquot and freeze for extended storage.
Catalog No. 50-001-84702 Supplier Medchemexpress LLC Supplier No. HYP7079210UG
Recombinant mouse placental growth factor (PLGF) expressed in HEK293 cells and provided lyophilized with a C-terminal His tag. PLGF binds VEGFR-1/FLT1 to stimulate angiogenesis and endothelial cell proliferation and migration; structurally it forms antiparallel homodimers and can heterodimerize with VEGF family members. Reconstitute according to the manufacturer instructions and aliquot for long-term storage.
  • Recombinant mouse PLGF expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Promotes angiogenesis via VEGFR-1/FLT1 binding.
  • Antiparallel homodimer that can form heterodimers with VEGF family members.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Approximate molecular weight 27-30 kDa by SDS-PAGE.
  • Store lyophilized at -20°C; aliquot and freeze for extended storage.