Learn More
Medchemexpress LLC PLGF Mouse HEK293 2ug
SDP
Catalog No. 5000197126
Encompass_Preferred
Recombinant mouse placental growth factor (PLGF) expressed in HEK293 cells with a His tag. Designed for research use in angiogenesis, endothelial cell proliferation and migration assays, and biochemical binding studies, this protein is provided at high purity for consistent experimental performance.
  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • His tag for straightforward purification and detection.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight approximately 27-30 kDa by SDS-PAGE.
  • Available in small, convenient sizes down to 2 μg for low-volume experiments.
Catalog No. 50-001-97126 Supplier Medchemexpress LLC Supplier No. HYP707922UG
Recombinant mouse placental growth factor (PLGF) expressed in HEK293 cells with a His tag. Designed for research use in angiogenesis, endothelial cell proliferation and migration assays, and biochemical binding studies, this protein is provided at high purity for consistent experimental performance.
  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • His tag for straightforward purification and detection.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight approximately 27-30 kDa by SDS-PAGE.
  • Available in small, convenient sizes down to 2 μg for low-volume experiments.