Learn More
Medchemexpress LLC PLGF Mouse HEK293 50ug
SDP
Catalog No. 5000184312
Encompass_Preferred
Recombinant mouse placental growth factor (PLGF) produced in HEK293 cells and provided as a purified His-tagged protein for in vitro research. Intended for applications in angiogenesis and endothelial cell biology, including ELISA, antibody validation, and cell-based signaling assays. Consult the manufacturer datasheet for handling, concentration, and storage instructions.
  • Expressed in HEK293 cells with a His tag.
  • Recombinant mouse PLGF for angiogenesis research.
  • Suitable for ELISA, antibody validation, and cell-based assays.
  • Supplied at 50 μg laboratory scale.
  • Purified and quality-tested; consult datasheet for lot-specific details.
Catalog No. 50-001-84312 Supplier Medchemexpress LLC Supplier No. HYP7079250UG
Recombinant mouse placental growth factor (PLGF) produced in HEK293 cells and provided as a purified His-tagged protein for in vitro research. Intended for applications in angiogenesis and endothelial cell biology, including ELISA, antibody validation, and cell-based signaling assays. Consult the manufacturer datasheet for handling, concentration, and storage instructions.
  • Expressed in HEK293 cells with a His tag.
  • Recombinant mouse PLGF for angiogenesis research.
  • Suitable for ELISA, antibody validation, and cell-based assays.
  • Supplied at 50 μg laboratory scale.
  • Purified and quality-tested; consult datasheet for lot-specific details.