Learn More
ACROBiosystems Recombinant Protein;Mouse PLGF / PGF Protein;HEK293;1MG;PGF,PGF,PLGF,PLGF1,PLGF-1,PlGF2,PlGF,PGFL,PlGF-2,SHGC-10760
Supplier:  ACROBiosystems PGFM52H01MG
Encompass_Preferred
Recombinant Protein;Mouse PLGF / PGF Protein;1MG;Mouse PLGF (PGF-M52H0) is expressed from human 293 cells (HEK293). It contains AA Val 19 - Pro 158 (Accession # P49764-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.8 kDa. The protein migrates as 27-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse VEGF R1, mouse IgG2a Fc Tag (Cat. No. VE1-M5256) at 2 μg/mL (100 μL/well) can bind Mouse PLGF (Cat. No. PGF-M52H0) with a linear range of 0.2-4 ng/mL (QC tested).
Catalog No. 50-165-7771