Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque PLGF / PGF Protein;HEK293;50UG;PGF,PGF,PLGF,PLGF1,PLGF-1,PlGF2,PlGF,PGFL,PlGF-2,SHGC-10760
Supplier:  ACROBiosystems PGFR52H050UG
Encompass_Preferred
Recombinant Protein;Rhesus macaque PLGF / PGF Protein;50UG;Rhesus macaque PLGF (PGF-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession # F7HB10-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 28-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) at 5 μg/mL (100 μL/well) can bind Rhesus macaque PLGF (Cat. No. PGF-R52H0) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-165-7960