Learn More
Medchemexpress LLC HY-A0122 1mg , Plicamycin CAS:18378-89-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA01221MG
Encompass_Preferred
HY-A0122 1mg Plicamycin CAS: 18378-89-7 Plicamycin is a selective specificity protein 1 (Sp1) inhibitor. Plicamycin inhibits the growth of various cancers by decreasing Sp1 protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9805