Learn More
Medchemexpress LLC Plicamycin (mithramycin A) | 18378-89-7 | MFCD00135618 | 99.6% | 10 MG
SDP
Catalog No. 5000298321
Encompass_Preferred
Plicamycin (mithramycin A) is an aureolic acid antibiotic and selective Sp1 transcription inhibitor used as a research reagent to modulate Sp1-dependent transcription and to study DNA/RNA synthesis interference. Supplied as a powder and in solution formats, it is provided in small research pack sizes and stored under low-temperature conditions.
  • Selective Sp1 transcription factor inhibitor for gene regulation studies.
  • Antineoplastic antibiotic that binds GC-rich DNA sequences to inhibit transcription.
  • Available in multiple small pack sizes and a 10 mM solution format in DMSO.
  • High reported purity (~99.6%), suitable for biochemical and cell-based assays.
  • Storage: powder at -20°C; solutions at -80°C for long-term stability.
Catalog No. 50-002-98321 Supplier Medchemexpress LLC Supplier No. HYA012210MG
Plicamycin (mithramycin A) is an aureolic acid antibiotic and selective Sp1 transcription inhibitor used as a research reagent to modulate Sp1-dependent transcription and to study DNA/RNA synthesis interference. Supplied as a powder and in solution formats, it is provided in small research pack sizes and stored under low-temperature conditions.
  • Selective Sp1 transcription factor inhibitor for gene regulation studies.
  • Antineoplastic antibiotic that binds GC-rich DNA sequences to inhibit transcription.
  • Available in multiple small pack sizes and a 10 mM solution format in DMSO.
  • High reported purity (~99.6%), suitable for biochemical and cell-based assays.
  • Storage: powder at -20°C; solutions at -80°C for long-term stability.