Learn More
Alkali Scientific Plicamycin [Mithramycin A], 5 Mg
SDP
Catalog No. 5000661606
Encompass_Preferred
Plicamycin (Mithramycin A), in a 5mg powder form, is an anticancer agent used primarily in chemotherapy for treating malignancies like testicular cancer and osteosarcoma. Its potent ability to inhibit DNA and RNA synthesis in cancer cells makes it an important tool in oncological therapies. In research, plicamycin is employed to explore gene regulation mechanisms and to inhibit cellular processes for studying cellular response to stress. This 5mg powder format provides precise dosing flexibility, essential for both clinical and laboratory settings. It is also used in antimicrobial research for its effectiveness against a variety of bacterial species, enhancing its applicability across multiple fields.
Catalog No. 50-006-61606 Supplier Alkali Scientific Supplier No. PLI002CP
Plicamycin (Mithramycin A), in a 5mg powder form, is an anticancer agent used primarily in chemotherapy for treating malignancies like testicular cancer and osteosarcoma. Its potent ability to inhibit DNA and RNA synthesis in cancer cells makes it an important tool in oncological therapies. In research, plicamycin is employed to explore gene regulation mechanisms and to inhibit cellular processes for studying cellular response to stress. This 5mg powder format provides precise dosing flexibility, essential for both clinical and laboratory settings. It is also used in antimicrobial research for its effectiveness against a variety of bacterial species, enhancing its applicability across multiple fields.