Learn More
Med Vet International Miltex College Cotton Pliers, w/Lock, Serrated, German, Each, 66-652
Catalog No. 502829179
Encompass_Preferred
Cotton pliers are used for handling cotton pellets, cotton rollers, small instruments, or other small items placed into or withdrawn from the mouth. The dressing forceps are also used to carry liquid medication between the closed beaks for deposit in area. Fitted with a lock, allowing the professional to keep things gripped with minimal effort. Serrated finish prevents slipping.
Catalog No. 50-282-9179 Supplier Med Vet International Supplier No. MIL66652
Cotton pliers are used for handling cotton pellets, cotton rollers, small instruments, or other small items placed into or withdrawn from the mouth. The dressing forceps are also used to carry liquid medication between the closed beaks for deposit in area. Fitted with a lock, allowing the professional to keep things gripped with minimal effort. Serrated finish prevents slipping.