Learn More
Chemglass Life Sciences De-Capper, Pliers, 20mm
SDP
Supplier:  Chemglass Life Sciences CG493021
Encompass_Preferred
De-Capper, Pliers, 20mm. Tool for decapping Aluminum seals on Microwave vials. The pliers decapper is an inexpensive way to remove seal caps by simply securing the seal between the jaws and squeezing the handles until cap pops off. For Microwave Vials and accessories, see CG-4920.
Catalog No. 50-872-710