Learn More
Med Vet International Miltex Light Wire Bird Beak Pliers, Forming, German, Each, 74-318
Catalog No. 502829005
Encompass_Preferred
Used for forming precise loops in light wire up to .020in.
Catalog No. 50-282-9005 Supplier Med Vet International Supplier No. MIL74318
Used for forming precise loops in light wire up to .020in.