Learn More
Med Vet International Miltex Utility Pliers, Bracket Remover, German, Each, 74-219
Supplier:  Med Vet International MIL74219
Encompass_Preferred
Precision grooved tips wedged between the bracket base and the tooth surface to safely and easily remove steel, ceramic and plastic brackets.
Catalog No. 50-282-8994