Learn More
TRIBIOSCIENCE INC TRIBIOSCIENCE INC Plain Pure POPC Liposomes (100 nm), 10 ML
SDP
Catalog No. NC3793934
Encompass
This preformed liposome reagent is prepared in 10% sucrose and contains NO drug loading battery. It is typically used as a placebo control for the drug loaded liposomes with the same lipid composition
Catalog No. NC3793934 Supplier TRIBIOSCIENCE INC Supplier No. F10110
This preformed liposome reagent is prepared in 10% sucrose and contains NO drug loading battery. It is typically used as a placebo control for the drug loaded liposomes with the same lipid composition