Learn More
enQuire Bioreagents Adipophilin / Perilipin-2 anti-Human Monoclonal [ADFP/1366], Size: 100ug
Supplier:  enQuire Bioreagents 123MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: ADFP/1366. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HepG2 or JAR cells. Liver, Adrenal gland or Cerebellum. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Adipophilin protein fragment (aa249-376). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 48kDa.Chromosome: 9p22.1. Gene Symbol: PLIN2. Gene ID: 123. Ensemble: ENSG00000147872. Uniprot: XP_016869748.
Catalog No. 50-188-4615