Learn More
Medchemexpress LLC HY-14444 5mg Medchemexpress, Plinabulin CAS:714272-27-2 Purity:>98%
SDP
Catalog No. 501932702
Encompass_Preferred
Medchemexpress, HY-14444 5mg Plinabulin CAS:714272-27-2 Plinabulin (NPI-2358) is a vascular disrupting agen ( VDA ) against tubulin-depolymerizing with an IC 50 of 9.8 nM against HT-29 cells [1] . Plinabulin binds the colchicine binding site of β-tubulin preventing polymerization and has potent inhibitory to tumor cells [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2702 Supplier Medchemexpress LLC Supplier No. HY144445MG
Medchemexpress, HY-14444 5mg Plinabulin CAS:714272-27-2 Plinabulin (NPI-2358) is a vascular disrupting agen ( VDA ) against tubulin-depolymerizing with an IC 50 of 9.8 nM against HT-29 cells [1] . Plinabulin binds the colchicine binding site of β-tubulin preventing polymerization and has potent inhibitory to tumor cells [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.