Learn More
Medchemexpress LLC HY-16050 1mg , Plitidepsin CAS:137219-37-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY160501MG
Encompass_Preferred
HY-16050 1mg Plitidepsin CAS: 137219-37-5 Plitidepsin (Aplidine) is a potent anti-cancer agent by targeting eEF1A2 ( KD=80 nM). Plitidepsin possesses antiviral activity and is against SARS-CoV-2 with an IC90 of 0.88 nM. Plitidepsin is usually used for multiple myeloma and advanced cancer research, and has the potential for COVID-19 research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9544