Learn More
Enzo Life Sciences PLK1 monoclonal antibody (3F8) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC105D
Encompass_Preferred
Clone: 3F8. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant fragment from the middle of human PLK1. UniProt ID: P53350. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Rat. Creossreactivity: Weakly cross-reacts with mouse Plk1. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7238