Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10044BA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Baculovirus. Species Reactivity: Human. Protein Expression: A DNA sequence encoding the Homo sapiens (Human) PLK1 / PLK-1, was expressed in the hosts and tags indicated. AA Sequence: See company website. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: PLK1. Gene ID: 5347. Uniprot: P53350.
Catalog No. 50-188-5483