Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PLK2, GST Tag Recombinant Protein

Catalog No. PV4204
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4204 10 μg
PR7592A 100 μg
PV4286 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV4204 Supplier Invitrogen™ Supplier No. PV4204
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

PLK2 is a serum inducible serine/threonine kinase that is a member of the polo family of serine/threonine kinases which have a role in normal cell division. The kinase may play a role in embryogenesis, wound healing, and neoplasia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NYY3
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 150mM NaCl, 0.5mM EDTA, 50% glycerol, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 10769
Molecular Weight (g/mol) 105.7 kDa
Name Human PLK2, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias h PLK-2; hPlk2; hSNK; Plk2; PLK-2; polo like kinase 2; polo-like kinase 1; polo-like kinase 2; Serine/threonine-protein kinase PLK2; serine/threonine-protein kinase SNK; Serum-inducible kinase; SNK
Common Name PLK2
Gene Symbol PLK2
Kinase Family PLK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
System Compatibility Z'-LYTE™
Target Entry PLK2
Target Gene polo-like kinase 2
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.