Learn More
Carnabio Usa Inc PLK3/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 051595
Encompass
PLK3, Catalytic domain, Wild type, Amino Acid 58-340, NP_004064.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0598