Learn More
Sino Biological Peptide Substrates: PLKtide
Catalog No. 502568238
Encompass_Preferred
The PLKtide peptide sequence (KKLGEDQAEEISDDLLEDSLSDEDE) is derived from the CDC25C protein sequence (182-204).
Catalog No. 50-256-8238 Supplier Sino Biological Supplier No. P4158
The PLKtide peptide sequence (KKLGEDQAEEISDDLLEDSLSDEDE) is derived from the CDC25C protein sequence (182-204).