Learn More
NANOSOFT POLYMERS PLL-g-PEG | 50mg
Catalog No. NC1728626
Encompass
PLL-g-PEG | Number of repeating units of PLL : X=200 | PEG MW: 5000 | Percentage of PEG substitution: 20% | 50mg
Catalog No. NC1728626 Supplier NANOSOFT POLYMERS Supplier No. 11354X20050002050MG
PLL-g-PEG | Number of repeating units of PLL : X=200 | PEG MW: 5000 | Percentage of PEG substitution: 20% | 50mg