Learn More
TARGETMOL CHEMICALS INC PLN-1474 5MG
SDP
Catalog No. 502579673
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PLN-1474 is a potent (alpha)V(beta)1 inibitor with IC50 < 50nM (WO2021127483 compound 1)purity: 99%
Catalog No. 50-257-9673 Supplier TARGETMOL CHEMICALS INC Supplier No. T601015MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PLN-1474 is a potent (alpha)V(beta)1 inibitor with IC50 < 50nM (WO2021127483 compound 1)purity: 99%