Learn More
Trajan Scientific PLNGR P10F-CTC-GT 2/PK
SDP
Catalog No. 502530282
Encompass_Preferred
Trajan Replacement Plungers, For Use With: For 10uL fixed needle gas tight syringe for CTC/Carlo Erba/ Fisons autosampler; Syringes 002987, 002991, IN VITRO use only. For lab analyzers only. Not for human use., Replacement Plunger, P10F-C/T-GT, PTFE Tip
Catalog No. 50-253-0282 Supplier Trajan Scientific Supplier No. 0318120
Trajan Replacement Plungers, For Use With: For 10uL fixed needle gas tight syringe for CTC/Carlo Erba/ Fisons autosampler; Syringes 002987, 002991, IN VITRO use only. For lab analyzers only. Not for human use., Replacement Plunger, P10F-C/T-GT, PTFE Tip