Learn More
Trajan Scientific PLNGR P25-CTC-GT
SDP
Catalog No. 502530281
Encompass_Preferred
Trajan Replacement Plungers, For Use With: For 25uL fixed needle gas tight syringe for CTC/Carlo Erba/ Fisons Autosampler; Syringe 003987, IN VITRO use only. For lab analyzers only. Not for human use., Replacement Plunger, P25F-C/T-GT, PTFE Tip
Catalog No. 50-253-0281 Supplier Trajan Scientific Supplier No. 031817
Trajan Replacement Plungers, For Use With: For 25uL fixed needle gas tight syringe for CTC/Carlo Erba/ Fisons Autosampler; Syringe 003987, IN VITRO use only. For lab analyzers only. Not for human use., Replacement Plunger, P25F-C/T-GT, PTFE Tip