Learn More
Cayman Chemical Plnt Aloe AloenIn 1mg
SDP
Catalog No. 502912484
Encompass_Preferred
A polyketide glucoside with diverse biological activities; an inhibitor of pancreatic lipase (IC50 = 14.95 UG/ml); an inhibitor of ADH and ALDH (IC50s = 33.1 and 27 µM, respectively, for the rat enzymes); active against antimony-sensitive L. donovani promastigotes (IC50 = 26 µM); inhibits gastric juice secretion in rats by 23% at 100 mg/kg
Catalog No. 50-291-2484 Supplier Cayman Chemical Supplier No. 358591 MG
A polyketide glucoside with diverse biological activities; an inhibitor of pancreatic lipase (IC50 = 14.95 UG/ml); an inhibitor of ADH and ALDH (IC50s = 33.1 and 27 µM, respectively, for the rat enzymes); active against antimony-sensitive L. donovani promastigotes (IC50 = 26 µM); inhibits gastric juice secretion in rats by 23% at 100 mg/kg